Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Isotemp™ Explosion-Proof Refrigerator/Freezers

For storage of volatile materials in areas with explosive atmospheres

Catalog No. 10ECEEFSA

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.

Attention:This item requires special delivery information. You will be asked for this during the checkout process.

Description

Description

Features:
  • Thermostat and compressor components are isolated within a vapor-proof enclosure
  • Electrical connections are hardwired to prevent sparking and accidental disconnection
  • Magnetic vinyl gaskets for positive door seal CFC-free insulation and refrigerant
  • Manual defrost

1 Length includes handles, height includes top-mounted controls.
2 Requires air clearance of 3 in. (7.6cm) around top and back, 1/2 in. (1.3cm) on sides.
3 Requires air clearance of 4 in. (10cm) around top, back, and sides.

Order Info

For 115V 60Hz. Not equipped with line cord and plug; must be hardwired into an appropriately rated conduit box to comply with NEC code.

Service Plan

Service agreement available.

Certifications

UL listed (two-door for Class 1 and Groups C and D hazard locations); CSA approved

Compliance

OSHA 29CFR1910.307

Warranty and Services

13 months, parts and labor

Specifications

Specifications

1°C to 12°C (Refrigerator), -12°C to -20°C (Freezer)
10.1 cu. ft.
115 V 60 Hz
1.2 A
28.6 x 23.63 x 59.75 in. (76.2 x 60 x 151.77 cm)
120 lb.
150 lb.
Temperature Display - None
ABS Plastic
White
Indoor use, Cold Storage Of Materials In Class I, Division I Group C and D, Class I Zone 1 Group IIB Environments
2 in. On Sides, 2 in. At Back, 2 in. On Top
Adjustable
10ECEEFSA
Automatic (Refrigerator), Manual (Freezer)
Explosion-Proof Freezer/Refrigerator
Hinged, Open Left to Right, Reversible
Polyurethane Foam
230 w
NFPA 45 and 70, UL
286 L
115 V
Not Provided
Ref: 18.25 x 19.75 x 38.25 in. (46.4 x 50.2 x 97.2 cm)Frz: 11.25 x 18 x 18.25 in. (28.6 x 45.7 x 46.3 cm)
54.5 kg
68 kg
R134a, 100% CFC Free
13 Months Parts and Labor
Enameled Steel
10 Years
Interior - No
2 Adjustable Feet In Front, 2 Fixed Pads In Rear
-12°C to -20°C
1°C to 12°C
Mechanical Dial Thermostat/Outside on Rear of Unit
Bottom Mounted
2 (2 compartments)
3 Adjustable Shelves, 1 Fixed Shelf, 1 Drawer, 4 Door Shelves
SDS
Documents
Product Certifications
Accessories
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit