Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Isotemp™ Flammable-Materials Storage Refrigerators: Basic Model

For reliable, safe storage of volatile materials, pharmaceuticals, clinical reagents, and biological samples

 View more versions of this product

Catalog No. 05FREEFSA

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.

Attention:This item requires special delivery information. You will be asked for this during the checkout process.

Description

Description

  • Adjustable temperatures from 1° to 12°C, depending on model
  • 50 cu. ft. (1420L) model features digital temperature display in 0.1° increments, °C or °F, and keyed power switches
  • Manual defrost
Safety Features
  • Orange warning label on front
  • Sparkproof interiors separate contents from compressor and motor
  • Compressors have enclosed overloads, relays, and thermostats that prevent sparks from escaping during door opening and closing
  • Static hot-wall condensers and evaporators are free of electric fans
  • Magnetic door gaskets provide airtight seals that insulate and prevent release of hazardous fumes
  • Top-mounted hydraulic thermostat and door stop
Construction
  • White, baked-on polyester enamel exteriors CFC-free polyurethane foam insulation and refrigerant
  • Solid door(s)

† Allow 3 in. (7.6cm) clearance top, back, and sides.
†† With legs.

Service Plan

Service agreement available.

Certifications

UL listed meets NFPA 45 and 70

Compliance

NFPA Code 99 (30 and 50 cu.ft. models only)

Warranty and Services

13 months, parts and labor

Specifications

Specifications

1°C to 12°C
5.5 cu. ft.
115 V 60 Hz
60 Hz
NEMA 5-15 P
17.5 x 20 x 27.7 in. (44.5 x 50.8 x 70.5 cm)
45.3 kg
54.5 kg
ABS Plastic
13 Months Parts and Labor
Enameled Steel
05FREEFSA
Flammable Material Storage Refrigerator
Hinged, Open Left to Right, Reversible
Polyurethane Foam
160 w
NFPA 45 and 70, UL
155.7 L
120 V
1.0 A
28.1 x 23.63 x 33.5 in. (71.37 x 64.8 x 85.1 cm)
100 lb.
120 lb.
R134a, 100% CFC Free
Door Lock
White
2 Adjustable Feet In Front, 2 Fixed Pads In Rear
Manual
Mechanical Dial Thermostat/Outside on Rear of Unit
Bottom Mounted
1 (1 compartment)
3 Adjustable Shelves, Storage On Bottom Of Unit
SDS
Documents
Product Certifications
Accessories
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit