Learn More
Fisherbrand™ Isotemp™ Value Lab Refrigerator
For safe, reliable storage of non-critical products.
 View more versions of this product
Catalog No. 17LREEFSA
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Description
Description
  • Factory-tested prior to shipment
  • NRTL-listed
  • Environmentally friendly, non-CFC refrigerants
  • Adjustable temperature control
  • Easy-to-clean, durable finish
  • Manual and auto defrost combination units
  • Manual defrost
Specifications
Specifications
1°C to 12°C
115 V
57.3 x 27.8 x 22 in. (145.4 x 70.49 x 55.8 cm)
5
Manual
16.6 cu. ft.
28.5 x 34 x 67.7 in. (72.4 x 86.4 x 171.9 cm)
White
17LREEFSA
4 fixed shelves, bottom of unit, 5 door shelves
SDS
Documents
Product Certifications
Accessories