Learn More
Fisher Scientific™ Isotemp™ Value Lab Refrigerators
For safe, reliable storage of non-critical products.
 View more versions of this product
Catalog No. 13-986-151B
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications
Description
  • Adjustable temperature control
  • Manual defrost
  • Adjustable shelves or baskets included
  • Environmentally friendly, CFC-free insulation and coolant
  • Factory tested prior to shipment
  • NRTL tested
  • Easy-to-clean, durable finish
Service Plan
Service agreement available.
Certifications
cCSAus
Warranty
13 months, parts and labor
Description & Specifications
Specifications
cCSAus
Mechanical Dial Thermostat/Inside, Lower Right Back Wall
6.5A
17 cu. ft.
2 Adjustable Feet In Front, 2 Fixed Pads In Rear
Bottom Mounted
Hinged, Opens Left to Right, Non-Reversible
28.5 x 34 x 67.7 in. (72.4 x 86.4 x 171.9cm)
60Hz
Polyurethane Foam
Yes/Three prong NEMA 5-15P, 3.048 Meters (10 Feet)
R134a
General Purpose Refrigerator
115 V/60 Hz
105kg
Isotemp™ general purpose laboratory refrigerator
White
5.0Amps
16.6 Cubic Feet/470.06 Liters
481.4L
Manual Defrost
1(1 compartment)
Epoxy Enameled Steel
22 x 27.8 x 57.3 in. (55.8 x 70.49 x 145.4cm)
ABS Plastic
17LREEFSA
4 Fixed Shelves, Bottom Of Unit, 5 Door Shelves
1° to 12°C
115V
230 lb.
13 Months Parts and Labor
Accessories