Learn More
Fisherbrand™ Isotemp™ Heating Blocks for Models 147Q and 145Q
For Isotemp™ Model 147Q and Model 145Q Dry-Bath Incubators
 View more versions of this product
Catalog No. 11-715-119Q
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Cast-aluminum alloy
  • Chemical resistant
  • Wells are 48mm deep
  • Dimensions: 3.75L x 3W x 2 in.H (9.5 x 7.4 x 51cm)
Warranty and Services
One year