Learn More
Fisher Scientific™ Isotemp™ Refrigerated/Heated Bath Circulators: 6.8-8.6L, 115V/60Hz Available on GSA/VA Contract for Federal Government customers only.
 View more versions of this product
Catalog No. 13-874-183
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
-35 to 100°C
1.79 to 2.27 gal.
115V
27.1 in.
20.8 in.
14.6 in.
±0.1°C
20.8 x 14.6 x 27.1 in. (52.8 x 37 x 68.8cm)
1kW
310mbar
17L/min
54.8kg
800w (at 20°C)
6.8 to 8.6L
60Hz
68.8cm
52.8cm
37cm
6.8 x 7.2 x 5.9 in. (W x L x D)
115V 60Hz
External circulation plumbing with 8 and 12mm fitting, supply and return clamps, work area cover
Force
121 lb.
4100 R35