Fisher Scientific™ Isotemp™ Refrigerator and Freezer Alarm Systems
For Isotemp and Isotemp Plus™ refrigerators and freezers
$1434.95
Specifications
For Use With Equipment Isotemp™ and Isotemp™ Plus™ refrigerator and freezer
View More Specs
Required Accessories: Remote systems require an existing alarm system, with dry alarm contacts, to be installed on the refrigerator or freezer in order to be compatible.
  • Products 2
  • Description & Specifications
Products
Catalog Number Product Type Price Quantity & Availability    
13-986-160
Freestanding Alarm System Each for $1,434.95
Check Availability
Add to Cart
13-986-100
Deluxe Remote System N/A N/A N/A  
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
Descriptions & Specifications
Related Products