Learn More
Fisherbrand™ Isotemp™ Shake Touch Shakers Accessory Blocks
Interchangeable blocks to accommodate a wide variety of tubes and microplates for use with Fisherbrand™ Isotemp™ Shake Touch shakers.
 View more versions of this product
Catalog No. 02-217-749
  Check Availability
Add to cart
Description
Description
Accessories Offered:
  • Well plate blocks with lid
  • PCR plate blocks with lid
  • Tube blocks
  • Conical tube blocks
  • Microplate thermal blocks
Specifications
Specifications
Tube block for 5 to 7mL tubes
3.43cm
Shake Touch Shakers
1.2cm
24
1.35 in.
5 to 7mL tubes
4.0 x 5.6 x 2.2” (10.2 x 14.2 x 5.6cm)
Heating/shaking
0.47 in.