Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Scientific™ Key Chain Laser Pointers

For slide presentations, microscope projections, field studies, wall charts and lectures

 View more versions of this product

Catalog No. 14-648-6

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Description

Key Chain Micro Laser Pointer

  • Half the size of pocket laser
  • Features output power of 4mW and range of 300 ft. (91m)
  • Brilliant red beam with 3.5mm diameter can be seen under normal lighting conditions
  • 2.37L x 0.56 in. dia. (6.0 x 1.4cm) body can be carried securely on keychain
  • Switch is momentary ON/OFF
Laser Key Chain
  • Brilliant red beam works equally well in daylight and dark
  • Easy to carry with attached key chain
  • Highly visible 650nm wavelength
  • Diameter, 3.5mm
  • Output, 4 mW Range, 400 ft.
  • Dimensions, 2.5 x 1 x 0.25 in. (6.35 x 2.54 x 0.64cm)
  • Weight, 0.5 oz. (14.2g)

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit