Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Scientific™ Lab Label Tape

Available in multiple colors

 View more versions of this product

Catalog No. 15-078-204


  Check Availability
Add to cart

Specifications

Description

Fisher Scientific™ Lab Label Tape can be unloaded and reloaded in less than 30 seconds.

  • Allows for color-coding labels
  • No batteries needed
  • Unit is always ready to use

  • Dimensions: 0.375 x 118 in. (0.90 x 300cm)

Description & Specifications

Specifications

Self-adhesive
Lab label maker
3m
0.010 x 3.277cm
Tape
4 Pack
Black
118 in.
0.375 x 118 in.
0.375 in.
Label Tape
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit