Learn More
Fisher Scientific™ Laboratory Homogenizer, Model 500
Efficient equipment with six generators (sold separately) available in Easy Care design with different diameters for various applications
 View more versions of this product
Catalog No. 08-451-667
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
0.1 to 4000mL
2.36 in.
50/60Hz
9.84 in.
75dBA
0° to 40°C
500w
6.5cm
230V 50/60Hz
Ideal for production of dispersions and emulsions, dispersion of plant, human or animal tissues, resins and pigments
6cm
Automatic soft start, integrated thermal overload fuse
25cm
32° to 104°F
230V
2.55 in.
9.84 x 2.55 x 2.36 in. (25 x 6.5 x 6cm)