Fisher Scientific™ MH Series Mechanical Heated Ultrasonic Cleaning Bath

For solid construction and excellent functionality, outfit your lab with a Fisher Scientific MH Series Heated Ultrasonic Cleaner.

 View more versions of this product

Catalog No. 15-337-433

  Check Availability
Add to Cart

    Specifications

    Dimensions (D x W x H) Interior 49.5 x 29 x 15cm (19.5 x 11.5 x 6 in.)
    Capacity 20.8L (5.5 gal.)
    Model M8800H
    Includes Tank drains with valves
    Material Plastic
    Timer 60 min.
    Voltage 120V

    • Sweep frequency feature eliminates standing waves and creates consistent cavitation throughout tank
    • Controls located above and behind cleaning tank in easily accessible control tower panel
    • Ultrasonic power tracking to track operating frequency of cleaner; always delivering the same power
    • Features 40kHz rugged industrial transducers
    • Chemical-resistant body made of plastic
    • Convenient built-in pour drains present on 0.5 gal. (1.9L) and 0.75 gal. (2.8L) models
    • Tank drains with valves incorporated on 1.5 gal (5.7L) and larger models
    • Includes 60-minute mechanical timer and continuous operation with heating capability
    • Tank cover included

  • Description & Specifications
  • Accessories
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit