Learn More
Fisherbrand™ Magnifier Lamp
2X magnification
Catalog No. 12-071-10
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Includes: Lamp and incandescent 60w bulb
Description
Description
Fisher Scientific™ Magnifier Lamp clamps vertically or horizontally to top or side of any edge.
  • Highly polished optical glass
  • Convenient front handle
  • 2X magnification
  • Sliding shutter cover protects lens when not in use
  • Large viewing area, 4 × 2.5 in. (10.2 × 6.4cm)
Compliance
UL listed and CSA approved
Specifications
Specifications
Magnifier Lamp, Swing Arm
Stand and lamp, 60w bulb
2x
Glass
60w Bulb
For lab and plant applications