Learn More
Fisherbrand™ FS6 Ultrasonic Cleaner
Fisher Scientific™ FS6 Ultrasonic Cleaner is compact and stylish, with convenience of plug-in-anywhere operation.
Catalog No. 15-335-6
Add to cart
Includes: Cover and parts basket
Description
Description
  • Five-minute timer shuts off automatically
  • Stainless-steel tank
  • Sealed within an impact-resistant plastic housing
Certifications
UL listed. CSA approved.
Specifications
Specifications
0.45
Plastic Housing, Stainless Steel Tank
Push bottom timer
120
22.3cm
120V 50/60Hz
6.5 in.
FS6
3 lb.
3.5 in.
15 oz
5 min.
40kHz
3.5 in.
6.5 in.l x 3.5 in.w x 2.25 in.d
Cleaning instrument and clock parts, small geological samples, small electrical/electronic components, metal and plastic machined parts, recorder and drafting pens
16.5cm
40w
1.36kg
8.89cm
Documents
Accessories