Fisher Scientific™ Micromaster™ Inverted Digital Microscopes with Infinity Optics
Infinity-corrected optics, with brightfield and phase contrast
 View more versions of this product
Catalog No. 12-563-518
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Height (English) 2.75 in.
    Includes Three vessel holders, 40X brightfield/phase infinity correct objective, X/Y axis attachable mechanical stage
    Voltage 115V
    Camera Type Digital
    Design Type Mechanical
    Magnification Power 40x, 100x, 200x, 400x
    Objective Types 40x, 100x, 200x, 400x
    For Use With (Application) Live cell observation
    Head Style Binocular
    Optical Design Infinity
    Warranty 2 Years
    These Micromaster inverted microscopes are ideal for live cell observation, featuring infinity-corrected optics, brightfield and phase contrast illumination techniques, with options for integrated digital camera with software.
    • Binocular, trinocular and digital options
    • Long working distance objectives
    • 2.75 in. working height (5.75 in. without condenser)
    • Infinity color corrected optical system
    Binocular (12-563-518)
    • Parfocal zoom
    • Magnification controls are dual, side-mounted knobs with clearly marked magnification values
    • Dual diopter adjustments allow for correction of dioptric differences
    • Interpuliary adjustment corrects for different spacing between eyes
    Trinocular (12-575-251 and 12-563-519)
    • 23mm photo port for attachment of camera
    • USB 2.0 direct connection to PC for image capture
    • View images through binocular and camera at the same time
    • Parfocality between camera and oculars
    • Micron USB2 software included
  • Description & Specifications
Related Products
Product Content Feedback