Fisher Scientific™ Micromaster™ Microscope Stage Micrometers
Used to calibrate eyepiece reticles for measurement
 View more versions of this product
Catalog No. 12-561-SM1
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    For Use With (Equipment) Micromaster™ microscope
    Product Type 1mm with 0.01mm division
    • Slide-mounted scales—also referred to as calibration plates
    • Fit any microscope
    Also available with Certificate of Traceability to NIST™ standards; contact your Fisher Scientific Customer Service Representative for information.
  • Description & Specifications
Related Products
Product Content Feedback