Learn More
Fisher Scientific™ Micromaster™ Microscope Stage Micrometers
Used to calibrate eyepiece reticles for measurement
 View more versions of this product
Catalog No. 12-561-SM3
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Micromaster™ microscope
2mm with 0.01mm division