Fisher Scientific™ Microplate Heaters

Designed for 96-well plate based cell-based and biochemical assays plus 3D cell culture in laminar flow hoods

 View all options for this product

Catalog No. 01-68R-08

 N/A
N/A

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Weight (Metric) 1.32kg
    Plug Type Switzerland
    Warranty 12 months

    The microplate heater maintains consistent temperature across its surface, ensuring cells in outer wells of microplate and inner wells are incubated at same temperature.

    • Robust construction
    • Achieve higher sensitivity and more rapid results with Thermo Scientific Pierce BCA protein assay kit
    • Two button temperature control (ambient +5°C to 65°C (ambient +9°F to 149°F))
    • Lightweight with small footprint only slightly bigger than standard microplate
    • Clear lid to maintain stable temperature during use in laminar flow hoods and for visibility
    • Smooth external surfaces for easy spraying and cleaning
    • Max. temperature set point: 65°C (149°F)
    • Min. temperature set point: 20°C (68°F)
    • Accuracy: ±1°C (±2°F)
    • Dimensions: 164 x 147 x135mm (6.4 x 5.7 x 5.3 in.)
    • Weight: 1.32kg (2.91 lb.)

    Applications:

    • Incubating biochemical assays
    • Incubating cell based assays
    • Trypsinising cells

    Warranty

    12 months

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit