Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Mini-Stirrer Products for Science Education

Large diameter top minimizes spill risks, making it ideal for student use.

Catalog No. S50450Q

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

 

  • Variable speed from 0 to 1650rpm
  • 6 dia. x 3 in. high (15.2 x 7.6cm) 
  • Front-mounted, stepless control knob
  • Strong aluminum top plate with baked enamel finish
  • Large stirring surface reduces spill risks
  • Includes stirring bars and support clamp (for mounting on lattice or other support)

 

Order Info

For 115V

Specifications

Specifications

0 to 1650rpm
115V
6 in.
15.2cm
SDS
Documents
Product Certifications
Accessories
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit