Learn More
Fisherbrand™ Model 120 Sonic Dismembrator
Programable system ideal for small volume liquid processing, 220V. Includes 1/8 in. probe for processing 0.5-15ml volumes. There are 3 probe size options available which allow for processing 0.2-50mL volumes.
 View more versions of this product
Catalog No. FB120A220
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Add to cart
Description
Description
Use this sonic dismembrator for effective analysis and sample preparation of small sample sizes. Fisher Scientific™ Model 120 Sonic Dismembrator offers both manual and programmable operation for consistent and reproducible results. Processes 0.2 to 50mL.
  • Processes 0.2 to 50mL
  • Programmable from 1 second to 10 hours
  • Overload protection
  • Pulse mode (1 second to 59 seconds)
Available accessories include:
Probes for individual samples
Eight tip horn for increased sample throughput
Cup Horn for indirect sonication
Lightweight converter for handheld operation
Specifications
Specifications
0.2 to 50mL
6 in.
50/60Hz
13.5 in.
1 sec. to 10 hrs.
120w
20.32cm
230V 50/60Hz, 120w
Cell disruption, protein extraction, DNA shearing/chip assay
15.24cm
Generator, converter, power cable, wrench set
34.29cm
230V
8 in.
13.5 x 8 x 6 in. (34.29 x 20.32 x 15.24cm)