Learn More
Fisherbrand™ Model 120 Sonic Dismembrator Available on GSA/VA Contract for Federal Government customers only.
Programable system ideal for small volume liquid processing, 220V. Includes 1/8 in. probe for processing 0.5-15ml volumes. There are 3 probe size options available which allow for processing 0.2-50mL volumes.
 View more versions of this product
Catalog No. FB120220
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Add to cart
Includes: Generator, converter, 1/8 in. probe, cables, wrench set
Description
Description
Use this sonic dismembrator for effective analysis and sample preparation of small sample sizes. Fisher Scientific™ Model 120 Sonic Dismembrator offers both manual and programmable operation for consistent and reproducible results. Processes 0.2 to 50mL.
  • Processes 0.2 to 50mL
  • 120 watts
  • Programmable from 1 second to 10 hours
  • Overload protection
  • Pulse mode (1 second to 59 seconds)
  • Energy Display(Watts and Joules)
  • Available accessories include: 3 Probe sizes for individual samples, Eight tip horn for increased sample throughput, Cup Horn for indirect sonication
  • Lightweight converter for handheld operation
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Order Info
Stand not included.
Specifications
Specifications
0.2 to 50mL
6 in.
50/60Hz
13.5 in.
FB120
220 to 230V
8 in.
13.5 x 8 x 6 in. (34.29 x 20.32 x 15.24cm)
20kHz (Ultrasonic Frequency)
Cell disruption, protein extraction, DNA shearing/ChIP assay
15.24cm
Generator, converter, 1/8 in. probe, power cable, wrench set. Note: Stand and Clamp shown in picture not included
34.29cm
1 sec. to 10 hrs.
120W
20.32cm
220 to 230V 50/60Hz
Documents
Accessories