Learn More
Fisherbrand™ Model 50 Sonic Dismembrator Available on GSA/VA Contract for Federal Government customers only.
Economical, small volume cell disruptor, 220V. Includes 1/8 in. probe for processing 0.5-15ml volumes. There are 3 probe size options available which allow for processing 0.2-50mL volumes.
 View more versions of this product
Catalog No. FB50A220
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Add to cart
Description
Description
The Fisher Scientific™ Model 50 Sonic Dismembrator is compact, portable and extremely simple to operate. Weighing less than 4 lb., this model is the smallest unit on the market and is highly effective for cell disruption, sample preparation and many other small volume applications.
  • Processes 0.2 to 50mL
  • 50 watts
  • Continuous or Remote operation (footswitch optional)
  • Lightweight converter for handheld operation
Basic cell disruption
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
0.2 to 50mL
5 in.
Generator, converter, power cable, wrench set
17.78cm
50w
20.32cm
230V 50/60Hz
Ideal for basic cell disruption
12.7cm
7 in.
230V
8 in.
7 x 8 x 5 in. (17.78 x 20.32 x 12.7cm)
20 kHz