Fisher Scientific™ Moticam™ Digital Cameras
Digital microscope camera with Motic software to create still or moving images on computer
 View more versions of this product
Catalog No. 15-000-110
  Check Availability
Add to Cart
    Specifications
    Warranty One year
    Pixels 2.8 x 2.8μm
    Resolution 1280 x 1024 (1.3MP) (WiFi only)
    Sensor Types CMOS
    Product Type Moticam X Digital Microscope Camera
  • Description & Specifications
Related Products
Product Content Feedback