Learn More
Fisherbrand™ OmniSette™ Processing/Embedding Cassettes
Just bend, snap apart, and lock
 View more versions of this product
Catalog No. 15-197-700C
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Disposable cassettes hold tissue specimens during embedding, processing, and storage in file
  • Molded from high-density polymer
  • Resist the effects of histological solvents and keep your specimens safely submerged
  • Efficient flow-through slots maximize fluid exchange and ensure proper drainage
  • Extra drains prevent trapped excess fluid
  • Reinforced ribs protect smaller specimens
  • One-piece integral lid eliminates need for separate steel lids and facilitates specimen handling
  • Lid snaps securely in place
  • Can be opened and closed as often as necessary during clearing, dehydrating, and infiltrating
  • Always relocks securely, with no danger of lost specimens
  • Color coding and large labeling area on three sides of cassette (20% larger than many other cassettes) ensures unmistakable identification with no increase in size†
Same capacity and displacement as most other commonly used cassettes
Specifications
Specifications
Healthcare