Learn More
Fisher Scientific™ Personal Microplate Shakers
Compact, economical portable shakers are ideal for any test environment including field applications
Alert:
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Includes: 15V 100mA DC power supply; Personal Plus adds clear plastic lid
Description
Counterbalanced orbital movement guarantees years of stable, quiet operation. Can be operated as stand-alone unit or integrated into a robotics system with remote on/off control (not included).
- Accommodate all microplate styles
- Momentary shaking action
- Variable speed control
- Maintenance-free, brushless DC motor
- Timer (Personal Plus only)
- Low-voltage power supply for added safety
- Washable anti-slip mat
- Operate as microtube shakers with accessory rack
Order Info
Microtube rack (03-692-196) available separately
Certifications
CE
Specifications
0.1 cm | |
120 V, 60 Hz | |
Personal Microplate shaker | |
50°F to 104°F | |
ABS | |
2.87 in. | |
5.25 in. | |
3.62 in. | |
1 to 180 min. |
950 to 1300 rpm | |
10°C to 40°C | |
CE | |
60 Hz | |
5.2 x 3.62 x 2.87 in. (133 x 9.3 x 7.2 cm) | |
7.2 cm | |
133 mm | |
9.3 cm |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.