Learn More
Fisher Scientific™ Polypropylene Controlled Heating Mantles
Features a durable, chemically resistant polypropylene outer housing. Fisher Scientific™ Polypropylene Controlled Heating Mantles minimize damage from spills and allow for maximum heat transfer with minimal risk of flask breakage.
 View more versions of this product
Catalog No. 11-474-550
Due to product restrictions, please to purchase or view availability for this product.
Specifications Description & Specifications
Specifications
250mL (8.4 oz.)
115V 50/60Hz, 150w
FEM0250/CEX1
200w
1.45cm
250mL
Polypropylene housing
842°F
450°C
0.57 in.
8.45 oz.
115V