Learn More
Fisher Scientific™ PowerGen Homogenizers Accessories: Saw Tooth Bottom Generator Probes Available on GSA/VA Contract for Federal Government customers only.
Saw teeth bottom stainless steel rotor stator probe
 View more versions of this product
Catalog No. 15-340-138
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Flat
0.25mL - 30mL
Flat bottom generator probe
316 L Stainless-steel
7mm
195mm
PowerGen homogenizers manufactured through 2013