Fisherbrand™ Precision Specific Gravity Hydrometers, 150mm Long Available on GSA/VA Contract for Federal Government customers only.
Requires 140mL liquid volume for submersion
 View more versions of this product
Catalog No. 11-556T
  Check Availability
Add to Cart
    Specifications
    Specific Gravity Range 1.72 to 1.79
    Increments 0.001
    Certifications/Compliance NIST, ASTM E100, ASTM E126
    Length (Metric) 165mm
  • Description & Specifications
Related Products
Product Content Feedback