Learn More
Fisherbrand™ Precision Specific Gravity 325 - 345mm Long Hydrometers
Requires 275mL liquid volume for submersion.
 View more versions of this product
Catalog No. 11-555AC
Add to cart
Description
Description
Requires 275mL liquid volume for submersion.
  • Use Cylinder 08-530K, available separately
  • Permanently marked with individual serial numbers
  • Accuracy meets or exceeds ASTM E 100 specifications
  • Calibrated according to ASTM E 126 specifications
  • With divisions of 0.001
  • Use Cylinder 08-530E, available separately
Order Info
A certificate of calibration is available; simply add the letter “C” to the end of the catalog number when ordering.
Certifications
A certificate is provided to indicate traceability to standards provided by NIST™.
Specifications
Specifications
Soda Lime Glass
3-point Certification, NIST, ASTM E100, ASTM E126
350mL Borosilicate Glass cylinder
0.0005
340mm
0.64 to 0.71