Learn More
Fisher Scientific™ Precision Specific Gravity Hydrometers, 340mm Long
Requires 275mL liquid volume for submersion
 View more versions of this product
Catalog No. 11-555NC
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
  • Use Cylinder 08-530K, available separately
  • Permanently marked with individual serial numbers
  • Accuracy meets or exceeds ASTM E 100 specifications
  • Calibrated according to ASTM E 126 specifications
  • With divisions of 0.001
  • Use Cylinder 08-530E, available separately
Description & Specifications
Specifications
0.0005
345mm
1.36 to 1.43
3-point Certification, NIST, ASTM E100, ASTM E126
350mL Borosilicate Glass cylinder