Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Refrigerator/Freezer Thermometer PROMO Products for Science Education

Insulated sensors protect against rapid temperature changes. Fisherbrand Refrigerator/Freezer Thermometer accurately monitor temperatures in freezers, water baths, heating blocks, incubators and refrigerators with bottle-enclosed “temperature buffered” sensor.

Catalog No. S66277PROMO

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Includes: AA battery, Velcro™ and magnetic strips for mounting of both thermometer and bottle sensor

Description

Description

  • Nontoxic glycol solution in bottle insulates sensor from rapid temperature changes when door is open
  • Triple display shows high, low and current temperatures, and min/max feature monitors high and low readings for any time period
  • Alarm feature provides an alert when temperature rises above or falls below a setpoint
  • 10 ft. micro-cable permits refrigerator doors to close on it
  • Velcro™ fasteners and magnetic strips are provided for mounting of both thermometer and bottle sensor
  • Range: -50° to +70°C (-58° to +158°F)
  • Resolution: 1°
  • Accuracy: ±1°C

Certifications

Individually serial-numbered Traceable™ Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™. It indicates traceability to standards provided by NIST™.

Specifications

Specifications

-58°F to +158°F
±1°C
158°F
−58°F
6.9 cm
4.25 x 2.75 in. (10.7 x 6.9 cm)
AA battery, Velcro™ and magnetic strips for mounting of both thermometer and bottle sensor
10.7 cm
4 oz.
2.75 in.
-50°C to +70°C
Digital LCD
70°C
−50°C
ISO 17025, A2LA, NIST
Monitoring liquids, air/gas or semi solids in freezers, water baths, outdoor tanks, incubators and refrigerators
4.25 in.
1°C
110 g
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit