Learn More
Fisher Scientific™ Replacement Tubing Set for Peristaltic Tubing Pumps
 View more versions of this product
Catalog No. 15-079-604
  Check Availability
Add to cart
Specifications
Description
  • 3/32-inch I.D. 4.25 in. Tubing Length
  • Silicone tubing and fittings/nipples (fitting color coded red inside)
  • For Cat. #3384, #3385 and #3386
Description & Specifications
Specifications
Silicone
-80° to 500°F
Yes
3/32 in.
Red
-62° to 260°C
Ultra-Low flow, Low flow and Medium flow Variable-Speed Peristaltic Tubing Pumps