Fisher Scientific™ Set of Eight Precision Specific Gravity Hydrometers Available on GSA/VA Contract for Federal Government customers only.

Permanently marked with individual serial numbers

 View more versions of this product

Catalog No. 11-582C

  Check Availability
Add to Cart

Alert

The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste.


    Specifications

    For Use With (Equipment) 300mL Borosilicate Glass cylinder
    Specific Gravity Range 0.695 to 2
    Temperature of Standardization 0 to 220°F
    Certifications/Compliance 3-point Certification, NIST, ASTM E100, ASTM E126
    Length (Metric) 300mm

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit