Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Spider Support Stands for Overhead Stirrers

Able to clamp on the edge of pails, drums, square or rectangular tanks.

 View more versions of this product

Catalog No. 14-512-911


Add to cart

Description

Description

Provides a safe and secure mounting system on any vessel

  • Fits securly on vessels with rims up to 5/8 in. wide
  • Adjustable legs and swivel clamp
  • Light weight aluminum base with a 5/8 in. diameter and 24 in. high stainless steel support rod

Specifications

Specifications

Vessel up to 15 in. inscribed circle
Spider Support Stand
1.58cm
71.2cm
Stainless Steel, Aluminum
0.62 in.
28 in.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit