Learn More
Fisherbrand™ Standard Exhaust Oil Filters
As supplied with Fisher Scientific Maxima C Plus pumps
 View more versions of this product
Catalog No. 01-257-5A
Add to cart
Description
Description
As supplied with Fisher Scientific Maxima C Plus pumps
These compact, round filters simply screw into threaded exhaust port to collect oil expelled during operation. A replaceable filter element within the oil filter separates the oil into particles no larger than 0.3µm, and drops collected oil back into the oil case.
  • Filter element is easily replaced
  • Reorder number located on filter canister
1For M4C or M8C vacuum pumps designed before September 1999 (S/N before FKK090000).
2For M4C or M8C vacuum pumps designed after August 1999 (S/N FKK090000 and newer).
Specifications
Specifications
63.5mm
Model M2C and M4C vacuum pump
6.35cm
Standard Exhaust Oil Filter
2.5 in.
2-1/2 in.