Learn More
Fisher Scientific™ Standard Grid-Top Adjustable-Flame Burners
Separate models for artificial and LP gases
 View more versions of this product
Catalog No. 03-902PQ
  Check Availability
Add to cart
Requires : 0.25 in. I.D. gas supply tubing recommended (not included)
Specifications Description & Specifications
Specifications
1.49 in.
Venturi tube for thorough mixing of air and gas, and adjustable air shutter and gas orifices for complete combustion
8.66 in.
LPG
3.8cm
2400 to 3300
22cm
Standard Grid Top Adjustable Flame Burner