Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BioTek™ Synergy™ Neo2 Multi-Mode Microplate Reader Available on GSA/VA Contract for Federal Government customers only.

Patented Hybrid Technology™ with independent filter and monochromator-based optics

$65250.00 - $125100.00

Specifications

Product Type Multi-Mode Reader
Absorbance Range 230 to 999 nm, 1 nm increments
Certifications/Compliance CE and TUV marked
Detector Type UV-Vis Absorbance, Fluorescence intensity, Luminescence, Fluorescence polarization, Time-resolved fluorescence, Alpha
Dynamic Range 0 to 4.0 OD
View More Specs

Products 8
Catalog Number Mfr. No. Includes Price Quantity & Availability  
Catalog Number Mfr. No. Includes Price Quantity & Availability  
BTNEO2 Greener Choice Available on GSA/VA Contract for Federal Government customers only.
Accessories View Documents
BioTek™
NEO2
UV-Vis absorbance, FI (Fluorescence Intensity)/FP (Fluorescence Polarization)/TRF (Time-Resolved Fluorescence), luminescence
Each for $65,250.00
Add to cart
 
BTNEO2M Greener Choice Available on GSA/VA Contract for Federal Government customers only.
Accessories View Documents
BioTek™
NEO2M
UV-Vis absorbance, top/bottom FI (Monochromator-based), top FI (Fluorescence Intensity)/FP (Fluorescence Polarization)/TRF (Time-Resolved Fluorescence), luminescence
Each for $91,850.00
Add to cart
 
NEO2MALPHA Greener Choice Available on GSA/VA Contract for Federal Government customers only.
Accessories View Documents
BioTek™
NEO2MALPHA
UV-Vis absorbance, top/ bottom FI, top FI (Fluorescence Intensity)/FP (Fluorescence Polarization)/TRF (Time-Resolved Fluorescence) (Filter), luminescence, top laser alpha
Each for $105,000.00
Add to cart
 
BTNEO2MB Greener Choice Available on GSA/VA Contract for Federal Government customers only.
Accessories View Documents
BioTek™
NEO2MB
UV-Vis absorbance, top/ bottom FI (Monochromator), top/bottom FI (Fluorescence Intensity)/FP (Fluorescence Polarization)/TRF (Time-Resolved Fluorescence), luminescence
Each for $110,050.00
Add to cart
 
NEO2MALPHAB Greener Choice Available on GSA/VA Contract for Federal Government customers only.
Accessories View Documents
BioTek™
NEO2MALPHAB
UV-Vis absorbance, top/ bottom FI, top/bottom FI (Fluorescence Intensity)/FP (Fluorescence Polarization)/TRF (Time-Resolved Fluorescence), luminescence, top alpa laser
Each for $125,100.00
Add to cart
 
BTNEO2ALPHA
Accessories View Documents
BioTek™
NEO2ALPHA
UV-Vis absorbance, FI (Fluorescence Intensity)/FP (Fluorescence Polarization)/TRF (Time-Resolved Fluorescence), luminescence, top laser alpha This product is not available for sale online. Please contact Customer Service at 1-800-766-7000 or send email to cs.webemail@thermofisher.com to discuss your specific assay requirements. N/A
BTNEO2B
Accessories View Documents
BioTek™
NEO2B
UV-Vis absorbance, top/bottom FI (Fluorescence Intensity)/FP (Fluorescence Polarization)/TRF (Time-Resolved Fluorescence), luminescence This product is not available for sale online. Please contact Customer Service at 1-800-766-7000 or send email to cs.webemail@thermofisher.com to discuss your specific assay requirements. N/A
NEO2ALPHAB
Accessories View Documents
BioTek™
NEO2ALPHAB
UV-Vis absorbance, top/ bottom FI (Fluorescence Intensity)/FP (Fluorescence Polarization)/TRF (Time-Resolved Fluorescence), luminescence, top laser alpha This product is not available for sale online. Please contact Customer Service at 1-800-766-7000 or send email to cs.webemail@thermofisher.com to discuss your specific assay requirements. N/A
Description

Description

  • Temperature Control – 4-Zone incubation to 70°C with Condensation Control™
  • Scientific quad monochromators with continuously variable bandwidth for optimal sensitivity
  • High performance filter system
  • Ultra-fast plate processing speeds with multiple PMT detectors
  • Live cell options: Controlled environment and direct bottom detection
  • Driven by best-in-class Gen5™ software for data collection and analysis
  • With Gen5 Secure Software
  • Includes dual top PMTs for fluorescence and luminescence

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Compliance

RoHS and 21 CFR Part 11

Specifications

Specifications

Multi-Mode Reader
CE and TUV marked
0 to 4.0 OD
HTS screeing, Drug absorption and metabolism, Biologics drug discovery and development, Drug discovery, Cell proliferation, Cytotoxicity, Biomarker quantification, Genetic analysis, Environmental testing, Food safety, Nuleic acid quantification, Protein quantification
≤1% from 0 to 3.0 OD
Linear, orbital, double orbital
±0.2°C at 37°C
39 cm
35 kg
230 to 999 nm, 1 nm increments
UV-Vis Absorbance, Fluorescence intensity, Luminescence, Fluorescence polarization, Time-resolved fluorescence, Alpha
250 W (max.)
Xenon Flash
0.0001 OD
Gen5™ Data Analysis Software, Gen5 Secure for 21 CFR Part 11 compliance (option)
52.5 cm
41 cm
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit