Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Tissue Culture Rotator
Requires : Tissue Culture Drum (order separately)
Description
Designed for growing tissue and virus cultures, hormone production, sample testing for the presence of viruses, and cytotoxicity assays.
- Variable rotation from 5 to 70rpm
- Elevation angle is adjustable from 5° above horizontal to 90°
- Drum can be lifted off roller without interrupting operation
- POWER-ON pilot light
- Can be used in standard incubators under controlled temperature conditions up to 40°C and humidity conditions from 20 to 80% noncondensing
- Powder-coated stainless-steel outer body is rugged and corrosion-resistant
- Long-life maintenance-free, brushless motor
Order Info
Order rotator body (14-251-250) and one drum for complete system. Dimensions (rotator body): 15D x 14W x 22.5 in. H (38 x 36 x 57cm). Unit weight (without roller): 19 lb. (9kg). All drums are 14 in. dia. (36cm) and weigh 5 lb. (2kg).
Warranty and Services
24 months
Specifications
12 to 70 rpm | |
120 V | |
60 W | |
Single-drum Tissue Culture Rotator | |
60 Hz | |
57.2 cm | |
38.1 cm | |
35.6 cm | |
9 kg | |
20 to 80%, Non-condensing |
120 V, 60 Hz | |
40°C (Max.) | |
0.5 A | |
cCSAus | |
22.5 in. | |
15 in. | |
14 in. | |
19 lb. | |
Designed for growing tissue and virus cultures, hormone production, sample testing for the presence of viruses and cytotoxicity assays |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction