Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Tissue Culture Rotator

Single-drum unit gently rotates up to 142 samples

 View more versions of this product

Catalog No. 14-251-253


Add to cart

Requires : Tissue Culture Drum (order separately)

Description

Description

Designed for growing tissue and virus cultures, hormone production, sample testing for the presence of viruses, and cytotoxicity assays.

  • Variable rotation from 5 to 70rpm
  • Elevation angle is adjustable from 5° above horizontal to 90°
  • Drum can be lifted off roller without interrupting operation
  • POWER-ON pilot light
  • Can be used in standard incubators under controlled temperature conditions up to 40°C and humidity conditions from 20 to 80% noncondensing
  • Powder-coated stainless-steel outer body is rugged and corrosion-resistant
  • Long-life maintenance-free, brushless motor

Order Info

Order rotator body (14-251-250) and one drum for complete system. Dimensions (rotator body): 15D x 14W x 22.5 in. H (38 x 36 x 57cm). Unit weight (without roller): 19 lb. (9kg). All drums are 14 in. dia. (36cm) and weigh 5 lb. (2kg).

Warranty and Services

24 months

Specifications

Specifications

14.17 in.
36cm
60 Tubes of 30mm dia.
Rotator drum
36cm
Designed for growing tissue and virus cultures, hormone production, sample testing for the presence of viruses and cytotoxicity assays
60.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit