Learn More
Fisherbrand™ Tissue Path™ IV Biopsy Cassettes
Finely perforated to prevent loss of small biopsy specimens during processing
Description
- Same two-piece molded hinge design as Tissue Path IV Cassettes
- Three-sided writing surface assures positive identification
- Resistant to solvents, decalcifying solutions and compatible with microwave techniques
Order Info
- Packaged in two dispenser boxes, 250 per box
Specifications
Powder Blue | |
Plastic | |
Biopsy Cassette |