Fisher Scientific™ Tissue Path™ Microsette™ Biopsy Cassettes
Innovative, disposable, plastic biopsy cassettes will make biopsy pads obsolete
 View more versions of this product
Catalog No. 15-182-706H
  Check Availability
Add to Cart
    Specifications
    Pore Style Mesh
    Material High Density Polymer
    Description Biopsy Cassette
    Writing Face Angle 45°
    Closure Type Integral Lid
    Color Gray
    No. of compartments Single Compartment
  • Description & Specifications
  • Accessories
Related Products
Product Content Feedback