Learn More
Fisherbrand™ Tissue Path™ Universal Modular Filing System
Stores 22% more slides or 32% more specimen blocks than standard units
 View more versions of this product
Catalog No. 22-244031
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • For microscope slides and histology cassettes
  • Made of extruded aluminum shells with a durable anodized finish
  • Slotted handle on each drawer for identification labeling
  • Interchangeable drawers
Specifications
Specifications
MicroSlide™ Drawer
Universal modular filing system