Learn More
Fisher Scientific™ Traceable™ Big-Digit Refrigerator Thermometer
Wide-range thermometer with 0.75 in. display for easy viewing
 View more versions of this product
Catalog No. 15-078-183
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
±1°C
3.75 x 1.5 x 0.75 in. (9.5 x 4.2 x 1.9cm)
0.75 in.
Lifelong Silver Oxide battery
9.5cm
22° to 140°F
2.5 oz.
4.2cm
ISO 17025, A2LA, NIST
Monitoring temperature in storerooms, labs, offices, hoods and outdoors
1.9cm
3.75 in.
0.1°C
30° to 60°C
1.5 in.