Learn More
Fisherbrand Traceable Countup Benchtop Timers
Handheld with a rugged ABS plastic case
$193.00
Catalog Number | Height (English) Display | Time Range | Weight (English) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Height (English) Display | Time Range | Weight (English) | Price | Quantity & Availability | ||||
06-662-40
|
0.75 in. | 10 hr. | 7 oz. |
Each for $193.00
|
|
06-662-1
|
0.5 in. | 100 hr. | 16 oz. |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
Fisher Scientific Traceable Countup Benchtop Timers provide single-action timing, time-in/time-out, and cumulative split functions. Handheld with a rugged plastic case.
- Battery is included
Certifications
Individually serial-numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA. It indicates traceability to standards provided by NIST.
Specifications
0.00003 | |
Rugged ABS Plastic case, battery |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.