Fisher Scientific™ Traceable™ Dual-Display Digital Stopwatch

Quartz-crystal design is accurate to 0.0005%—over 100 times more accurate than other stopwatches

 View more versions of this product

Catalog No. 14-648

  Check Availability
Add to Cart

Includes: Stopwatch; lanyard

    Specifications

    Dimensions (L x W x H) 2.5 x 3 x 0.75 in. (6.35 x 7.62 x 1.9cm)
    Display Type LCD
    Height (English) Display 0.25 in.
    Height (English) 0.75 in.
    Height (Metric) 1.9cm
    Includes Stopwatch; lanyard; Battery
    Length (English) 2.5 in.
    Length (Metric) 6.35cm
    Resolution 0.01 sec.
    Weight (English) 2.5 oz.
    Weight (Metric) 71g
    Width (English) 3 in.
    Width (Metric) 7.62cm
    Product Type Dual-Display Digital Stopwatch
    Time Range 10 hrs.

    Engineered for ease of operation, accuracy, and reliability. Fisher Scientific™ Traceable™ Dual-Display Digital Stopwatches provide precise, Traceable timing for enzyme tests, viscosity measurements, animal studies, and all timed lab tests. They are made for scientists in industrial, educational, hospital, and governmental labs. Quartz-crystal design is accurate to 0.0005%—over 100 times more accurate than other stopwatches.

    Set countdown time from 10 hours to 1 second. When zero is reached, timer alarms and repeats countdown from the programmed time. Repeat cycle automatically continues until stopped. Display count box shows how many times the cycle has been repeated. LCD screen continually displays time remaining to zero. 

    • Functions include single-action, time-out, interval-split, cumulative split, continuous roll-over timing, and countdown alarm timing
    • Dual display shows cumulative and interval split
    • Features eight recallable memories, plus fastest and slowest lap times
    • Repeat timer provides repeat function of countdown alarm timing
    • Tough, water-resistant ABS plastic case seals out moisture, dust, and fumes encountered in harsh lab, plant, or field environments
    • Timing range is 9 hours, 59 minutes, 59 seconds, and 99 hundredths
    • Resolution is 0.01 second
    • Accuracy is 0.0005%
    • Size: 6.4 x 7.6 x 1.9cm (2.5 x 3 x 0.75 in.)
    • Weight: 2.5 oz.

    Order Info

    Replacement battery: Cat. No. 02-440-1A

    Certifications

    Individually serial-numbered Traceable™ Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA. It indicates traceability to standards provided by NIST.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit