Fisher Scientific™ Traceable™ International Standards Extreme-Accuracy Digital Thermometers

Ideal for measuring temperatures in water baths, cuvets, gas systems, reagents, standards solutions, ovens, incubators, pharmaceuticals, petroleum products, foods, and wastewater

 View more versions of this product

Catalog No. 15-078-180

  Check Availability
Add to Cart

Includes: Stainless Steel probe, High-impact, chemical-resistant ABS plastic case, stand, Velcro™, magnetic strips, wall mount, two AA batteries (allow continuous, 1-year always-on monitoring)

    Specifications

    Temperature Range (English) 37.00° F
    Temperature Range (Metric) 35.00 to 39.00°C,37°C
    Accuracy ±0.05°C
    Diameter (English) Probe 0.12 in.
    Dimensions (L x W x H) 4.25 x 2.75 x 0.75 in. (10.7 x 6.9 x 1.9cm)
    Display LED,LED
    For Use With (Application) Measuring temperatures in water baths, cuvets, gas systems, reagents, standards solutions, ovens, incubators, pharmaceuticals, petroleum products, foods and wastewater
    Height (English) 0.75 in.
    Height (Metric) 1.9cm
    Includes Stainless Steel probe, High-impact, chemical-resistant ABS plastic case, stand, Velcro™, magnetic strips, wall mount, two AA batteries (allow continuous, 1-year always-on monitoring)
    Length (English) 4.25 in.
    Length (English) Probe 6.25 in.
    Length (Metric) 10.7cm
    Length (Metric) Probe 15.9cm
    Material ABS Plastic,ABS Plastic
    Resolution 0.01°C
    Weight (English) 5 oz.
    Weight (Metric) 140g
    Width (English) 2.75 in.
    Width (Metric) 6.9cm
    Certifications/Compliance ISO 17025, A2LA, NIST
    For Use With Measuring temperatures in water baths, cuvets, gas systems, reagents, standards solutions, ovens, incubators, pharmaceuticals, petroleum products, foods and wastewater

    Ideal for measuring temperatures in water baths, cuvets, gas systems, reagents, standards solutions, ovens, incubators, pharmaceuticals, petroleum products, foods, and wastewater. Fisher Scientific™ Traceable™ International Standards Extreme-Accuracy Digital Thermometers have an impervious stainless-steel probe that shows slightest temperature change in liquid, air/gas, and semisolids.

    Units read 0.00, 25.00, 37.00, or a combination of all three in °F and °C.

    • Provides accurate readings with tip penetration of 0.33 in.
    • Records time/date for min/max readings and hi/lo alarm alerts
    • Displays minimum/maximum and current temperatures
    • Min/max monitors readings over any period; displays exact time/date of min/max temperature
    • Alarm provides two visual LEDs and two audio alerts when temperature exceeds set points; displays exact time/date of dual thermometer alarms
    • Programmable in 0.1° increments; alarms signal continuously even if temperature returns to non-alarm range
    • Resolution: 0.01°
    • Accuracy: ±0.05°C within ±2°C of standard
    • Probe Dia.: 0.12 in. (0.30cm)
    • Stem Length: 6.25 in. (15.87cm)
    • Overall Length: 9 in. (22.86cm)
    • Cable: 5 ft. (1.5m)

    Certifications

    Individually serial-numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™; indicates traceability to standards provided by NIST™.

    Compliance

    Meets ITS-90, International Temperature Standard.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit