Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Traceable™ Jumbo-Digit Radio-Controlled Wall Clock
SCI_ED

Jumbo sized 3.75 in. clock digits are readable from up to 100 ft. away.

Catalog No. S04708

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »

This item is not returnable. View return policy

Description

Description

Design allows mounting on any wall or standing on any work surface.

  • Compact case measures only 9W × 5 3/4 in.
  • Radio signal from NIST™'s Colorado atomic clock automatically sets correct date and time
  • Calendar displays month and date and day of the week
  • Thermometer displays ambient temperature in °F and °C
  • Clock has 12 or 24-hour time-of-day display
  • Resolution: 1 second
  • Accuracy: 1/10,000 second when receiving radio signal, otherwise 0.01%
  • Temperature range: 23 to 122°F (-5 to 50°C)
  • Resolution: 0.1°
  • Accuracy: ±1°C

Certifications

Individually serial-numbered Traceable™ Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™.

Specifications

Specifications

Jumbo Digit Compact Radio Controlled Wall Clock
1 sec.
-5°C to +50°C
0.1°
5 in.
9 in.
1.25 in.
15 oz.
Wall mount, bench stand, batteries and Traceable Certificate
Each
0.0001 sec.
23°F to 122°F
±1°C
5 x 9 x 1.25 in. (12.7 x 22.8 x 3.17 cm)
12.7 cm
22.8 cm
3.17 cm
0.42 kg
Design allows mounting on any wall or standing on any work surface
ISO 17025, A2LA, NIST
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit