Fisher Scientific™ Traceable™ Logger-Trac™ Temperature Datalogger Accessories
Accessories for Traceable™ Logger-Trac™ Temperature Datalogger
 View more versions of this product
Catalog No. 15-081-148
  Check Availability
Add to Cart
    Specifications
    For Use With (Equipment) Fisher Scientific Traceable Logger-Trac Temperature Datalogger
    Length (Metric) 1.5m
    For Use With (Application) Transfer recorded information in dataloggers to computer
    Type Cradle/USB cable
    Certifications/Compliance RoHS compliant
    Accessories for Traceable™ Logger-Trac™ Temperature Datalogger
    15-081-145
    • Waterproof paper sleeves
    • For protecting temperature probe sensor
    • Transparent
    15-081-148
    • USB standard A to Mini B, IC-9P or USB to serial converter
    • USB Version 2.0, ELV and RoHS compliant
    • Cable length: 1.5m
    • Multiple cradles allow for batch programming up to 8 units
  • Description & Specifications
Related Products
Product Content Feedback