Learn More
Fisherbrand™ Traceable™ Memory Monitoring Thermometer Replacement Probe
Replacement probe for Traceable™ Memory Monitoring Thermometer
Catalog No. 15-079-602
Add to cart
Description
Description
Replacement probe for Traceable™ Memory Monitoring Thermometer
  • Probe without bottle
  • Probe is 5/16in. in diameter and 3/4in.long
  • Ten foot cable
  • For use with 15-077-8D
Specifications
Specifications
0.31 in.
10 ft.
Replacement Probe
10 ft. cable
0.75 in.