Fisher Scientific™ Traceable™ Platinum Ultra-Accurate Digital Thermometer

High-precision unit is designed for years of reliable service—even in the severest environments

 View more versions of this product

Catalog No. 15-081-103

  Check Availability
Add to Cart

Includes: Probe, cable, Plastic carrying case, traceable certificate and 9V battery; Computer output supplied with unit

    Specifications

    Temperature Range (Metric)  -100.000° to +200.000°C,-100.00 to +200.000°
    Includes Platinum probe, cable, computer output capabilities (software package must be purchased separately), plastic carrying case, batteries and Traceable Certificate

    Accuracy ±0.05°C
    Diameter (English) Probe 0.25 in.
    Dimensions (L x W x H) 11.5 x 3.5 x 5.5 in. (29.21 x 8.89 x 13.97cm)
    Display LCD,LCD
    For Use With (Application) Measuring temperatures in gas systems, reagents, chemical solutions, soil, water baths, ovens, incubators, pharmaceuticals, foods and wastewater
    Height (English) 5.5 in.
    Height (Metric) 13.97cm
    Length (English) 11.5 in.
    Length (English) Cable 120 in.
    Length (English) Probe 1 in.
    Length (Metric) 29.21cm
    Length (Metric) Cable 304.8cm
    Length (Metric) Probe 2.54cm
    Resolution 0.0001°C
    Weight (English) 8 oz.
    Weight (Metric) 226.7g
    Width (English) 3.5 in.
    Width (Metric) 8.89cm
    Certifications/Compliance ISO 17025, A2LA, NIST
    Depth (English) 1.12 in.
    Depth (Metric) 2.84cm
    For Use With Measuring temperatures in gas systems, reagents, chemical solutions, soil, water baths, ovens, incubators, pharmaceuticals, foods and wastewater

    High-precision unit is designed for years of reliable service—even in the severest environments. Fisher Scientific™ Traceable™ Platinum Ultra-Accurate Digital Thermometers have platinum sensor which ensures linear response and high accuracy across the entire range. Ideal for foods, pharmaceuticals, petroleum products, wastewater, gas systems, soil, ovens and electronics manufacturing.

    • Handle Probe: Meter range of -200° to +500°C with 0.001° or 0.0001° resolution
    • Bullet Probe: Meter range of -100 to 200°C
    • Displays °F or °C
    • Membrane-covered keys are splash-proof and easy to operate
    • Decimal-placement key changes display to show one, two, three, or four significant numbers after decimal
    • Internal data logger automatically captures MIN/MAX readings every hour for last 24 hours
    • Instant response probe updates readings three times per second for liquids, air/gas, semisolids
    • HOLD allows user to freeze display for reading later

     

    Handle Probe Model

    • Range: -99° to +500°C
    • Fast-response 4-wire platinum waterproof stainless steel probe
    • Probe diameter: 0.13 in.
    • Stem length: 6.30 in.
    • Overall length: 9 in.
    • Cable length 5 ft.
    • Waterproof
    • Use for liquids, air/gas and semisolids with readings at tip penetration of 0.3 in.
    Bullet™ Probe Model
    • Range: -99° to +500°C
    • Fast-response 4-wire platinum waterproof stainless steel Bullet probe
    • Probe diameter: 0.25 x 1 in.
    • With high temperature (500°C), 10 in. flexible glass cable
    • Waterproof
    • Use for liquids, air/gas, and semisolids

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit